Learn More
Thermo Scientific™ Frames and Accessories for Immuno Breakable Modules, BreakApart frame
Use Thermo Scientific™ Frames and Accessories for Immuno Breakable Modules with Nunc™ C8 Breakable Modules.
Brand: Thermo Scientific™ 465404
62.60 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Code : 76
Additional Details : Weight : 1.37000kg
Description
Use Thermo Scientific™ Frames and Accessories for Immuno Breakable Modules with Nunc™ C8 Breakable Modules. These frames include tools to facilitate removing the strips.
Warranty: 90 daysSpecifications
Strip Plate | |
10 Pk., 60 Cases | |
Frame | |
C | |
60 | |
Immuno Standard Modules |
High optical quality for reliable results | |
White | |
ABS Plastic | |
For solid phase immuno assays | |
5 | |
60/Cs. |