Learn More
Tocris Bioscience™ FRAX 486
Potent PAK inhibitor; brain penetrant and orally bioavailable
Brand:  Tocris Bioscience™ 5190/10
159.82 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18711678
  • £206.00 / 10mg
Explore more special offers
Description
Description
FRAX 486 is a potent p21-activated kinase (PAK) inhibitor (IC50 values are 14, 33, 39 and 575 nM for PAK1, PAK2, PAK3 and PAK4 respectively). Blocks and reverses the DISC1 knockdown-induced reduction in dendritic spine size in cortical neurons. Attenuates dendritic spine elimination and enhances spine generation in DISC1 knockdown mice. Ameliorates autism-like behavioral symptoms in fragile X mental retardation 1 (Fmr1) knockout mice. Brain penetrant and orally bioavailable.
Specifications
Specifications
>98%
FRAX 486
FRAX 486
10 mg
513.39