missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ FreeStyle™ MAX Reagent
Click to view available options
Quantity:
1 mL
15 mL
10 x 15 mL
Unit Size:
15mL
1mL
Pack of 10
Description
Includes
Each unit contains one vial of reagent.
- Allows highest expression levels and transfection rates with lowest cytotoxicity in bio-production applications
- Formulated for use with:
- FreeStyle 293-F Cells (suspension human embryonal kidney cells, Cat. No. R790-07) in serum-free FreeStyle 293 Expression Medium (Cat. No. 12338-018)
- FreeStyle CHO-S Cells (suspension Chinese Hamster Ovary cells, Cat. No. R800-07) in serum-free FreeStyle CHO Expression Medium (Cat. No. 12651-014)
- DG44 Cells (DHFR– suspension CHO cells) in CD DG44 Medium with 8 mM L-glutamine and 18 mL/L 10% Pluronic F-68 (Cat. No. A11000-01)
- FreeStyle™ MAX Reagent is a proprietary, animal origin–free formulation for transfecting plasmid DNA into eukaryotic cells that can easily be scaled up to produce large amounts of recombinant proteins
- Intended for use with the FreeStyle™ MAX 293 Expression System (Cat. No. K9000-10) and the FreeStyle™ MAX CHO Expression System (Cat. No. K9000-20).
Order Info
Shipping Condition: Wet Ice
Specifications
Specifications
| Classification | Animal Origin-Free |
| Content And Storage | Each unit contains one vial of reagent. Store at +4°C. FreeStyle™ MAX is guaranteed stable for 6 months when stored properly. |
| Cell Type | Established Cell Lines |
| Sample Type | Plasmid DNA |
| Transfection Technique | Lipid-based Transfection |
| For Use With (Application) | Transfection |
| High-throughput Compatibility | Not High-throughput Compatible (Manual) |
| Product Line | Freestyle |
| Product Type | Transfection Reagent |
| Quantity | 1 mL |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction