Learn More
Welch™ Freeze Dryer Adapter Kit 3
Shop All Welch Ilmvac Products
Click to view available options
For Use With (Equipment):
Freeze Dryer
Unit Size:
Each
Description
- Allows connection of various types of laboratory glassware to the freeze dryer system
- Connection: DN 16 KF
- Includes different adapters and connectors allowing for easy attachment of flasks, vials, ampules or other glassware
- PVC Construction
- Enables efficient and controlled freeze drying of samples
- Enhances versatility and functionality
Specifications
Specifications
| For Use With (Equipment) | Freeze Dryer |
Documents
Scientific Resources
Brochures
Fisherbrand Vacuum pumps
File size: 5.6 MB
Product Content Correction
Product Title
Welch™ Freeze Dryer Adapter Kit 3 >
Spot an opportunity for improvement?Share a Content Correction