Learn More
Gardner Denver Welch™ Kit 5 Freeze dryer adapter kit, for Chemvac & Vacuum concentrator Adapter kit, DN 16 KF
For rotary vane pump DN 16 KF
Brand: Gardner Denver Welch™
Code : NEW
Additional Details : Weight : 0.29000kg
Description
Vacuum concentrator kit includes:
- KF16 flange adapter
- 1.5m PVC vacuum hose
- Clamps, centring rings and adapters
Specifications
DN 16 KF | |
Rotary vane pump DN 16 KF |