missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Freeze Drying Beaker Flasks and Accessories

Freeze drying beaker flasks and accessories for use with a lyophilizer.

Brand:  BUCHI 11066143

 View more versions of this product

Product Code. 17244674

  • £286.00 / Each
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For use with a lyophilizer
Specifications

Specifications

1200 mL
Freeze Dryers
200 mm
3.7 lb.
Freeze Drying
Beaker Flask
-80°C to +400°C
1.7 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers