Learn More
Azpack™ Freeze Drying Stoppers
Push in to engage and pull out to remove stopper. Azpack™ Freeze Drying Stoppers are nonthreaded and constructed of sturdy rubber.
Brand: Azpack™ 10483274
Description
- For use with Freeze Drying and Injection Vials
- Nonthreaded push-in stopper
- Use with appropriate polypropylene screw thread cap
Specifications
Bromobutyl Rubber | |
500 Pack | |
2mL glass vials | |
13 mm |
Product Content Correction
Product Title
Azpack™ Freeze Drying Stoppers > Diameter (Metric): 13mm; For 2mL vials
Spot an opportunity for improvement?Share a Content Correction