missing translation for 'onlineSavingsMsg'
Learn More

Labcold Freezer RLBF1740/LK 50L super spark free, bench top -30°C to -40°C

Brand:  Labcold RLBF1740/LK

Freezer RLBF1740/LK 50L super spark free, bench top -30°C to -40°C

Code : 50

Additional Details : Weight : 0.01000kg

Product Code. 11598974

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers