missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ FreeZone™ Stoppering Tray Dryers, Isolation Valve, 230V Models

Provides precise control and program flexibility for consistent performance. Labconco™ FreeZone™ Stoppering Tray Dryers, Isolation Valve, 230V Models are designed for lyophilizing moderate to large loads and stoppering vials or serum bottles under vacuum. They offer options for built-in 6-port manifold and isolation valve.

£33955.00

Specifications

Type Stoppering Tray Dryer
No. of Shelves 3
Temperature Range (Metric) -40°C to +40°C
Material Steel
Certifications/Compliance UL, CAN/CSA, CE
View More Specs

Requires: Freeze dryer, vacuum pump


Products 1
Product Code Brand Includes Plug Type Price Quantity & Availability  
Product Code Brand Includes Plug Type Price Quantity & Availability  
15772867
View Documents
Labconco™
794801250
3 shelves, Isolation Valve British (UK)
£33955.00
Each
Estimated Shipment: 04-07-2025
to see stock.
Add to basket
 
Description
Show More
Specifications

Specifications

Stoppering Tray Dryer
-40°C to +40°C
UL, CAN/CSA, CE
24.6 in.
32.4 in.
27.1 in.
9A
60 Hz
195.49 kg
3
Steel
3 shelves, Isolation Valve
62.4 cm
82.2 cm
68.8 cm
230 V
431 lb.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Labconco™ FreeZone™ Stoppering Tray Dryers, Isolation Valve, 230V Models

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.