missing translation for 'onlineSavingsMsg'
Learn More

Vileda Professional™ Roll-O-Matic™ Sponge Mop Refills

Vileda Professional Roll-O-Matic™ sponge mop refills are made in the USA with “Advanced Technology” foam. They are manufactured specifically for the controlled environment industry, and allow 25% more liquid absorption and 2X more liquid release than competing foams.

Brand:  Vileda Professional™ 119616

Additional Details : Weight : 2.04000kg

 View more versions of this product

Product Code. 17842456

  • £126.27 / Pack of 12
Estimated Shipment: 29-08-2024
to see stock.
Add to basket

Description

Description

Vileda Professional Roll-O-Matic™ sponge mop refills are made in the USA with “Advanced Technology” foam. They are manufactured specifically for the controlled environment industry, and allow 25% more liquid absorption and 2X more liquid release than competing foams.

All Vileda Professional brand refills are ideal for use with the "Pull and Lift" technique most commonly used in controlled environments. They are ideal for general use, absorbing spills, applying disinfectant, and cleaning particulates.

  • Fits all original and self-wringing Roll-O-Matic™ mops
  • 10- or 14-inch options for both stainless steel and galvanized steel handles
  • Completely autoclavable and resistant to gamma and ETO sterilization
  • Individually packaged and lot number controlled
  • Certificate of conformance and technical data available upon request
  • Sterile versions validated to 10-6 per ISO 11137, individually double bagged with lot number and indicator dot, and supplied with certificate of sterility upon request
Specifications

Specifications

Roller Mop Refill
Non-sterile
10 in.
Original and Self Wringing Roll-O-matic Roller Mops
For Tear Free Cleaning of Deep Contaminants and Disinfectant Residues
Blue, White
Autoclavable
12/Cs.
“Disinfect”
Polyester, Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers