Learn More
Vileda Professional™ Swep Duo Plus Mop Frame
For use in all sectors and building types. Vileda Professional Swep Duo Plus Mop Frame is suitable for both horizontal and vertical surfaces, and is efficient even in corners and other hard-to-reach spaces.
Brand:  Vileda Professional™ 137878
Product Code. 17882476
  • £352.00 / Pack of 10
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Built-in lever designed to allow hands free removal of used mop heads
  • Double sided that works with all core and specialist application
  • Reinforced swivel joint part
  • Trapezoidal shape for easy cleaning of corners
  • Attaches to extendable handles (Recommended Aluminum telescopic handle - 119966)
  • Lightweight and ergonomic
Specifications
Specifications
Mop Frame
50 cm (19.7 in.)
Floors, Walls and Ceilings, ISO 5, LAR, Production, Labs
Autoclavable
Telescopic Handle, Aluminum Handle