Learn More
IKA™ Fritte Set
This Fritte Set contains two glass frits for the Pro-Divide reaction cell.
Brand:  IKA™ 0020102634
Additional Details : Weight : 0.00100kg
 View more versions of this product
Product Code. 17583626
  • £66.00 / Pack of 2
Estimated Shipment: 05-11-2024
to see stock.
Add to basket
Description
Description
  • This set contains two glass frits for the Pro-Divide reaction cell.
  • The size of each pore falls between these ranges:
    • 1 μm set: 1 and 1.6 μm
    • 5 μm set: 4 and 5.5 μm
    • 10 μm set: 10 and 16 μm
Specifications
Specifications
Fritte Set
10 μm
Pro-Divide Reaction Cell