missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Frizzled-9 Antibody (vantictumab) - IgG2SA, Novus Biologicals™

Human Monoclonal Antibody

£157.00 - £674.00

Specifications

Antigen Frizzled-9
Clone vantictumab
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30047304
View Documents
Novus Biologicals™
NBP3-28265-1mg
1 mg N/A N/A N/A
30044615
View Documents
Novus Biologicals™
NBP3-28265-50ug
50 μg
£157.00
50µg
Please to purchase this item. Need a web account? Register with us today!
30046607
View Documents
Novus Biologicals™
NBP3-28265-100ug
100 μg
£249.00
100µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Frizzled-9 Monoclonal antibody specifically detects Frizzled-9 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications

Specifications

Frizzled-9
Flow Cytometry, ELISA, Functional Assay
Unconjugated
Human
GPCR, Growth and Development, Neuronal Cell Markers, Signal Transduction, Wnt Signaling Pathway
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
8326
IgG2
Protein A purified
Store at -20°C in powder form. Store at -80°C once reconstituted.
vantictumab
Monoclonal
Purified
RUO
Human
CD349, CD349 antigen, frizzled (Drosophila) homolog 9, frizzled homolog 9 (Drosophila), frizzled-9, fz-9, FZD3fzE6, FzE6, hFz9
FZD
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Novus Biologicals™ Frizzled-9 Antibody (vantictumab) - IgG2SA, Novus Biologicals™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.