Learn More
3M™ Front-Fitting Reusable Full Face Mask
3M™ Reusable Full Face Mask features a panoramic visor that allows for a virtually unrestricted field of vision. Compatible with the 3M™ Filters, DT Series, this Full Face Mask is suitable for a wide range of wearers and applications.
£1390.00 - £1645.00
Product Code | Brand | Size | Model | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Model | Price | Quantity & Availability | ||||
17736993
|
3M™
7100265075 |
Small | FF-601F |
£1645.00
Pack of 4 |
Estimated Shipment: 09-09-2024
Log in to see stock. |
||||
17726993
|
3M™
7100265074 |
Medium | FF-602F |
£1390.00
Pack of 4 |
Estimated Shipment: 09-09-2024
Log in to see stock. |
||||
17766993
|
3M™
7100265236 |
Large | FF-603F |
£1645.00
Pack of 4 |
Estimated Shipment: 09-09-2024
Log in to see stock. |
||||
Description
- Panoramic visor to allow for a virtually unrestricted field of vision
- Visor is designed to minimise visual distortion
- Soft, highly flexible and conforming Liquid Silicone Rubber faceseal to enhance user comfort
- Broad faceseal, designed to minimise pressure points and provide an effective seal without over-tightening the head harness
- Matt finish TPE inner mask helps to prevent glare on the inside of the visor
- 4/Cs.
Specifications
Full Face Respirator |