Learn More
OHAUS™ Frontier™ 5000 Series Micro Centrifuge
OHAUS™ Frontier™ 5000 Series Micro Centrifuge provides high-speed operation for a range of essential lab applications. Engineered with convenient operation in mind, these centrifuges feature an intuitive interface, making all key parameters immediately accessible for observation or adjustment.
Brand:  OHAUS™ 30370691
1676.75 GBP valid until 2025-12-31
Use promo code "24319" to get your promotional price.
 View more versions of this product
Product Code. 16221742
  • £1765.00 / 1 unit
Explore more special offers
Description
Description
  • Excellent functionality when used with rotors compatible with 1.5/2.0 mL or 5 mL microtubes with high throughput and biocontainment options, spin columns, PCR strips, cryo tubes and hematocrit capillaries.
  • Designed for ease of operation, Frontier Micro centrifuges feature convenient controls on an intuitive interface, allowing responsive operation – even when wearing gloves.
  • Packaging all their functionality within a small footprint, Frontier Micro centrifuges save valuable space on the lab bench.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Backlit LCD, 2 lines
230 V
FC5513+R01
Yes
13.100 kg
Microcentrifuge
356.00 mm
200 to 13,500 rpm
High sample throughput,multi-safety control, personal program function
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
OHAUS™ Frontier™ 5000 Series Micro Centrifuge > Rotor Included: Yes