Learn More
Frosters Cryogenic Apron
Product Code. 15127965
Click to view available options
Size:
Large
Specifications
Type Cryogenic Apron
Size Large
Material Polyolefin, Polyester, Cotton, Polyamide
Color Blue
Resistance Water, spills, and splashes of Cryogenic material
For Use With (Application) The apron provides optimum protection for torso and upper legs against the effects of spills and splashes of cryogenic material
Dimensions (L x W) 1200 x 620mm
Length (Metric) 1200 mm
Width (Metric) 620 mm