missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fructosyl amino acid oxidase Antibody, HRP, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NBP1-77782H
Additional Details : Weight : 0.00970kg
Description
Fructosyl amino acid oxidase Polyclonal antibody specifically detects Fructosyl amino acid oxidase in Bacteria samples. It is validated for Western Blot, ELISASpecifications
Fructosyl amino acid oxidase | |
Polyclonal | |
Western Blot, ELISA | |
EC=1.5.3.-, FAOX-C | |
Fructosyl amino acid oxidase [from Corynebacterium sp. Expressed in E.coli] (Uniprot: Q8RIU8) | |
0.1 mL | |
Primary | |
Store at 4C in the dark. |
Western Blot, ELISA | |
HRP | |
PBS | |
Sheep | |
Multi-step | |
RUO | |
Bacteria | |
IgG |