missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fructosyl amino acid oxidase Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Bio-Techne NBP1-77782-0.1MG
Additional Details : Weight : 0.00970kg
Description
Fructosyl amino acid oxidase Polyclonal specifically detects Fructosyl amino acid oxidase in Bacteria samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Spécification
Fructosyl amino acid oxidase | |
Polyclonal | |
Western Blot 1:500 - 1:5000, ELISA 1:5000 - 1:20000, Immunoprecipitation 1:100 | |
Q8RIU8 | |
Sheep | |
Ion exchange chromatography | |
RUO | |
Bacteria | |
Purified |
Western Blot, ELISA, Immunoprecipitation | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
faoxC | |
Fructosyl-Amino Acid Oxidase [from Corynebacterium sp. Expressed in E.coli] | |
0.1 mg | |
Primary | |
Store at 4C. Do not freeze. | |
IgG |