Learn More
Thermo Scientific Chemicals FSB Buffer
For frozen storage of competent cells
Brand: Thermo Scientific Chemicals J60262.=E
Code : NEW
Additional Details : Weight : 0.00012kg
Description
For frozen storage of competent cells
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Colorless | |
Frozen storage buffer | |
Liquid | |
Buffer |
125 mL | |
2°C to 8°C | |
For frozen storage of competent cells |
RUO – Research Use Only