missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FSD1L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21032-1-AP-150UL
This item is not returnable.
View return policy
Description
FSD1L is a protein coding gene.Specifications
FSD1L | |
Polyclonal | |
Unconjugated | |
FSD1L | |
CCDC10, CSDUFD1, FSD1 like protein, FSD1 N terminal like protein, FSD1CL, FSD1L, FSD1NL, MIR1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
313208, 319636, 83856 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BYN5, Q9BXM9 | |
Fsd1l | |
FSD1L Fusion Protein Ag15345 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |