Learn More
Specac™ FT-IR Gas Pack Complete
Brand:  Specac™ GS01170
Code : Z3
Additional Details : Weight : 0.01000kg
Product Code. 15259817
  • £920.81 / Each
Estimated Shipment: 09-08-2024
to see stock.
Description
Description
The Storm 10cm Pyrex gas Cell is central to this Pack and is ideal for measuring the infrared spectra of gases, gas mixtures and vapours. Also includes KBr windows, seals and a cell mount
Specifications
Specifications
KBr windows, seals and a cell mount