Learn More
Specac™ FT-IR Research Starter Kit
Brand:  Specac™ GS01190
Encompass_Preferred
Product Code. 15209827
  • £6843.76 / 1 set
Explore more special offers
Description
Description
This kit includes a Quest ATR ZnSe accessory with liquid cell , KBR + CaF2 windows, spacers, nujol, fluorolube, luer syringe, 2 ton mini-pellet press, 7mm die, pellet ring holder, pestle and mortar, KBr powder and mount
Specifications
Specifications
A Quest ATR ZnSe accessory with liquid cell , KBR + CaF2 windows, spacers, nujol, fluorolube, luer syringe, 2 ton mini-pellet press, 7 mm die, pellet ring holder, pestle and mortar, KBr powder and mount