Learn More
Fuel Cell Car Science Kit
Combines cutting-edge science, education, and fun for all
Brand: Edulab
Code : 50
Additional Details : Weight : 1.50000kg
Includes: Car, solar panel, water-filling syringe, all necessary components, instruction manual and a Renewable Energy Education CD
Description
- Uses a reversible PEM fuel cell that combines electrolysis and power conversion into one device
- Oxygen and hydrogen gases are formed in two transparent water containers
- Steers independently of the user once in operation
- Runs past obstacles; if the car hits a barrier, it automatically reverses 90°
- Grades: 6+
Renewable Energy Education CD
- Renewable Energy Science Education Manual: Introduces renewable energy in seven chapters—Global Warming and Climate Change, Solar Technologies, Wind Technologies, Electrolyzers, Fuel Cells, Hydrogen Storage, and Basic Electronics
- Experiment Manual: More than 20 experiments for middle to high school level sciences in several sections—Solar Energy, Wind Energy, Energy from Hydrogen (Fuel Cell), and “Ultra Cool« experiments
- Flash Animations: Describe and show how a fuel cell works, different types of fuel cells, electrolysis, and various applications for fuel cells
- Assembly Guides
- Technical Support with videos
Specifications
Fuel Cell Car Science Kit | |
Demonstrates electrolysis of water into oxygen and hydrogen | |
Car, solar panel, water-filling syringe, all necessary components, instruction manual and a renewable energy education CD |
6+ | |
Electrolysis and power conversion into one single device |