Learn More
3M™ Fuel™ Safety Glasses
Click to view available options
Lens Tint:
Blue, Mirror
Bronze
Indoor/Outdoor, Mirror
Red, Mirror
Frame Color:
Bronze
Silver
Unit Size:
Pack of 20
Description
- Safety glasses designed to be worn at work, at home or during outdoor recreational activities
- The frame's molded lens supports prevent the lenses from being dislodged by a strong frontal impact
- Ventilation system on nose bridge and temples to help prevent fogging
Specifications
Specifications
| Certifications/Compliance | CE Approved, EN 166:2001 |
| Lens Coating | Scratch-resistant |
| Description | Blue Mirror coating for increased durability and improved vision |
| Lens Tint | Blue, Mirror |
| Product Type | Safety Glasses |
| Frame Color | Silver |
| Lens Material | Polycarbonate |