missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Full Coverage Face Shields with Anti-Fog
Shop All SP Scienceware Products
Click to view available options
Bel-Art™ Full Coverage Face Shields with Anti-Fog
Description
- Large, wrap-around design provides coverage of entire face with unrestricted visibility
- Helps protect the face from potential hazards such as dust, dirt, pathogens, sprays and fluids
- Anti-fog coating on interior surface prevents fogging for clear vision
- Comfortable; suited for extended wear
- Thick top foam guard provides distance between user and shield
- Clear PET plastic viewing area; remove protective film prior to use
- Disposable shield; single use only
- Full Coverage Face Shields with Anti-Fog have not been FDA cleared or approved
- The product has been authorized by FDA under an EUA for use by healthcare providers as personal protective equipment.
- This product is only authorized for the duration of the declaration that circumstances justifying the authorization of emergency use under Section 564(b)(1) of the Act, 21 USC 360bbb-3(b)(1) unless the authorization is terminated or revoked sooner.
Specifications
Specifications
Certifications/Compliance | CE |
Height (English) | 9 in. |
Height (Metric) | 23 cm |
Thickness (Metric) | 0.36 mm |
Width (English) | 13 in. |
Width (Metric) | 33 cm |
Lens Coating | Anti-fog |
Lens Tint | None |
Product Type | Full Coverage Faceshield |
Quantity | 20/Pk. |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Documents
Scientific Resources
Brochures
CE_17145811.pdf
File size: 871.8 kB
Product Information
Product Guides
CE Certificate
File size: 870 kB
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bel-Art™ Full Coverage Face Shields with Anti-Fog >
Spot an opportunity for improvement?Share a Content Correction