missing translation for 'onlineSavingsMsg'
Learn More

MULTIROIR Full Eurobox Container

Product Code. p-7217333
Click to view available options
Dimensions (L x W x H):
400 x 300 x 117 mm
400 x 300 x 120 mm
400 x 300 x 220 mm
400 x 300 x 290 mm
600 x 400 x 100 mm
600 x 400 x 220 mm
600 x 400 x 320 mm
600 x 400 x 420 mm
Color:
Blue
Gray
Red
White
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17236890

Brand: MULTIROIR BA12001

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Full Eurobox container with solid bottoms and solid walls, ideal for the storage and transport of goods

  • Full Eurobox container
  • The best quality / price ratio on the market
  • Solid bottoms and solid walls
  • Ideal for the storage and transport of goods
  • Stackable and high load capacity
  • Temperature resistant from -20 ° C to + 110 ° C

Specifications

Type Storage Box
Material Polypropylene
Color Gray
Shape Rectangular
Dimensions (L x W x H) 400 x 300 x 117 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.