missing translation for 'onlineSavingsMsg'
Learn More

CleanSpace™ Full Face Mask Orinasal Inner Mask Set for PAPR Assembly

CleanSpace™ Full Face Mask Orinasals designed to be used with the CleanSpace™ Full Face Mask are made of silicone and polycarbonate.

Brand:  CleanSpace™ PAF-1022

 View more versions of this product

Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15920092

  • £121.00 / Each
Estimated Shipment: 27-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Used with the revolutionary CleanSpace™ full face mask
  • Designed for comfort over long periods
  • Allows breathability and prevents fogging
  • Easy to wash and quick drying
  • Designed for long wear in harsh environments
  • Easy and quick replacement
  • Cleaning: Machine washable or use in warm soapy water. Do not use solvents (turpentine or acetone), hot water, bleaching or chemical agents
Specifications

Specifications

AS/NZS1716: 2012, AS/ NZS 1337.1:2010 High Impact Resistance
EN 12942
Small
Welding, woodworking, manufacturing, smelting, construction, recycling plants, emergency services, mining, agriculture, processing plants, grinding
PAF-1022
120 x 80 x 90 mm
EN 12942
PAPR Full Face Mask
PAPR full face mask only
PAF-1014, PAF-1106
Store at -10°C to 55°C
5.5 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers