Learn More
Kimberly-Clark™ KleenGuard™ A40 Arm Covers, White, One Size Product Code.: 12982245
In Stock
to see stock.
Add to basket
Kimberly-Clark™ KleenGuard™ A40 Arm Covers, White, One Size
Product Code. 12982245
White
Click to view available options
Color:
White
Product Code. 12982245
Brand: Kimberly-Clark™ 98730
In Stock
to see stock.
Edge
Add to basket
Add to basket
Single-use arm covers constructed using the A40 anti-static base sheet, offering enhanced protection for body parts that are more exposed to contamination. Provides hygiene and comfort in low-risk areas (CE class 1) and prevents damage to clothes and can help to reduce costs.
Ideal for:
  • Keeping clothes clean
  • Helping to maintain hygiene and reduce contamination in non-hazardous, food processing and light industrial areas without leaving lint
  • Wearing over protective clothing to help them last longer in heavier industrial areas.
Available as: crisp, white, breathable, non-absorbent one-size sleeves with elasticated cuffs.
  • One-size, white sleeves with elasticated cuff (200 total)
  • Single-use lint free non-absorbent light duty sleeve
  • Keep clothing clean and reduce contamination in food processing and light industrial areas
  • Provides additional protection
  • PPE Category I - CE Simple
Specifications
Size Universal
Product Type General Purpose Sleeve
Material Microporous Film Laminate
Color White
Disposable Single-use
No. per Pack 200 Pk.
Product Content Correction
Product Title
Kimberly-Clark™ KleenGuard™ A40 Arm Covers, White, One Size >