missing translation for 'onlineSavingsMsg'
Learn More

Sonics Materials Full Wave Solid Probe

Product Code. 11744328
Click to view available options
Diameter (Metric):
13 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11744328

Brand: Sonics Materials 6300217

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

This high intensity, microprocessor controlled, ultrasonic processor features a full wave solid probe ½ in. (13mm) solid – 10 in. (254mm) long.

  • Full wave solid probe ½ in. (13mm) solid – 10 in. (254mm) long
  • High intensity ultrasonic processor
  • Microprocessor controlled
  • 600-Watt model

Specifications

Product Type Full Wave Solid Probe
Diameter (Metric) 13 mm
Length (Metric) 254 mm
Packaging Quantity 1 Ea.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.