Learn More
Fulvestrant, MedChemExpress
MedChemExpress Fulvestrant (ICI 182780) is a pure antiestrogen and a potent estrogen receptor (ER) antagonist with an IC50 of 9.4 nM. Fulvestrant is also a GPR30 agonist. Fulvestrant effectively inhibits the growth of ER-positive MCF-7 cells with an IC50 of 0.29 nM. Fulvestrant also induces autophagy and has antitumor efficacy.
Brand: Medchem Express HY-13636
Description
Fulvestrant (ICI 182780) is a pure antiestrogen and a potent estrogen receptor (ER) antagonist with an IC50 of 9.4 nM. Fulvestrant is also a GPR30 agonist. Fulvestrant effectively inhibits the growth of ER-positive MCF-7 cells with an IC50 of 0.29 nM. Fulvestrant also induces autophagy and has antitumor efficacy.Specifications
Fulvestrant | |
White | |
C32H47F5O3S | |
ICI 182780 ZD 9238 ZM 182780 | |
C[C@@]12[C@@H](O)CC[C@@]1([H])[C@]3([H])[C@H](CCCCCCCCCS(CCCC(F)(F)C(F)(F)F)=O)CC4=C(C=CC(O)=C4)[C@@]3([H])CC2 | |
606.77 | |
Research | |
Cancer-programmed cell death |
129453-61-8 | |
Research | |
10 mg | |
DMSO : 250 mg/mL (412.02 mM; Need ultrasonic) | |
606.77 | |
95.0% | |
Solid |