missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Mycotoxin produced by Fusarium moniliforme. Potently inhibits sphingosine N-acyltransferase (ceramide synthase), causing an accumulation of sphingoid bases (IC50 ∽ 75 nM). Also inhibits protein phosphatases; IC50 values are 80, 300, 400, 500 and 3000 μM for PP5, PP2Cα, PP2A, PP1γ2 and PP2B respectively.

Specifications
Specifications
CAS | 116355-83-0 |
Molecular Formula | C34H59NO15 |
Quantity | 1 mg |
Synonym | macrofusine, fumonisin b1, 2r,2'r-1,2,3-propanetricarboxylic acid 1,1'-1s,2r-1-2s,4r,9r,11s,12s-12-amino-4,9,11-trihydroxy-2-methyltridecyl-2-1r-1-methylpentyl-1,2-ethanediyl ester, 2s-2-2-5r,6r,7s,9s,11r,16r,18s,19s-19-amino-6-3r-3,4-dicarboxybutanoyl oxy-11,16,18-trihydroxy-5,9-dimethylicosan-7-yl oxy-2-oxoethyl butanedioic acid |
InChI Key | UVBUBMSSQKOIBE-UWCNXKJPSA-N |
SMILES | CCCCC(C)C(C(CC(C)CC(CCCCC(CC(C(C)N)O)O)O)OC(=O)CC(CC(=O)O)C(=O)O)OC(=O)CC(CC(=O)O)C(=O)O |
IUPAC Name | (2S)-2-[2-[(5R,6R,7S,9S,11R,16R,18S,19S)-19-amino-6-[(3R)-3,4-dicarboxybutanoyl]oxy-11,16,18-trihydroxy-5,9-dimethylicosan-7-yl]oxy-2-oxoethyl]butanedioic acid |
Molecular Weight (g/mol) | 721.838 |
PubChem CID | 16760030 |
Formula Weight | 721.83 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fumonisin B1, Tocris Bioscience™ >
Spot an opportunity for improvement?Share a Content Correction