missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ FunctionMAX™ TiterEnhancer
GREENER_CHOICE

Product Code. p-7075914
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 13436426

Brand: Gibco™ A1501001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

FunctionMAX™ TiterEnhancer

FunctionMAX™ TiterEnhancer uses a new technology to combine key nutrients in a highly concentrated (150 g/L), pH-neutral, chemically-defined, AOF (animal origin-free) functional additive that is designed to reinforce existing feeds and amplify the productivity of standard high cell density fed-batch platforms. FunctionMAX™ TiterEnhancer is available in both this liquid format and an AGT™ dry format and is stable for over 1 year. FunctionMAX™ TiterEnhancer offers:

  • Up to double prior fed-batch titers
  • Highly concentrated ingredients
  • Easy to use pH-neutral format
  • Specialized delivery of ingredients found in base media—nothing exotic
  • Use that is economical

    When to Choose FunctionMAX™ TiterEnhancer:

  • If you have a drop off in specific productivity late in a high density fed-batch culture, FunctionMAX™ TiterEnhancer may be able to improve your titers
  • If you have an extremely low peak cell density fed-batch culture, you may want to consult with PD-Direct™ Services for advice on cell adaptation and/or fed-batch optimization
  • If you have no drop off in specific productivity late in a high density fed-batch culture and still need higher titers, you may want to consult with PD-Direct™ Services for advice on fed-batch optimization

    Up to Double Prior Fed-batch Titers
    We have demonstrated 35–120% product titer improvements when FunctionMAX™ TiterEnhancer was added on top of existing processes or as a partial feed replacement (Figures 1–3), resulting in a lower total proportion of feed addition. These increases were achieved primarily by maintaining specific productivity throughout each fed-batch process. Improvements were seen immediately and without process optimization. We have also shown that low-volume addition of FunctionMAX™ TiterEnhancer was sufficient to improve titers for several processes using different commercially available media and feeds, with those having higher cell densities seeing the most benefit. This additive is not designed to completely replace other feeding supplements that provide additional ingredients not found in FunctionMAX™ TiterEnhancer.

    Highly Concentrated Ingredients
    The technology used to deliver this functional additive promises to transform recombinant drug bioprocessing by raising key ingredient concentrations to levels that are out of reach using conventional approaches. A concentration of 150 g/L is achieved with FunctionMAX™ TiterEnhancer (compared to 20–25 g/L for most basal media), and it has been shown to be stable in dry or liquid format for over 1 year.

    Easy-to-Use pH-neutral Format
    Reconstitution of the AGT™ dry format is surprisingly simple (add water and stir), with all components going into solution. There is no pH adjustment required, and being pH neutral it is safer in handling and use than extreme pH alternatives. As a pH neutral additive, this product will help avoid bioreactor complications often seen with extreme pH supplements. Further, the liquid is very stable—so much so that the liquid format version is made from the same AGT™ dry format material that is also available for purchase. The liquid is provided for small-scale trials and the AGT™ dry format for seamless scale-up in use.

    Specialized Delivery of Ingredients Found in Base Media—Nothing Exotic
    FunctionMAX™ TiterEnhancer does not introduce new ingredients to your fed-batch process; it simply provides the same key ingredients found in basal media that the cells need late in culture to maintain their productivity. Further, the technology used to combine these ingredients does not introduce any additional ingredients. Nor does it contain any growth factors or undefined components like hydrolysates. A DMF (drug master file) reference can be requested for this product for assurance of regulatory authorities that there are no substances or ingredients in the formulation that might create regulatory concerns.

    Economies in Use
    FunctionMAX™ TiterEnhancer is designed to deliver highly concentrated and high value addition to your fed-batch process. Consider that it provides 7-fold the ingredient concentration of a basal medium and can provide up to a doubling of titer. Further, in normal use at about 10% of working volume, the actual price per liter of use is similar to that of basal media.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage 100 mL Gibco™ Bottle
    Cell Line CHO
    Classification Animal Origin-free, Chemically-defined, Protein-free
    Form Liquid
    Product Type Enhancer
    Serum Level Serum-Free
    Sterility Sterile-filtered
    Without Additives No Glutamine
    Product Line FunctionMAX™ TiterEnhancer
    Quantity 100 mL
    Shipping Condition Room Temperature
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.