missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAT™ Safety Funnel 'ARNOLD' with Ball and Lid, V2.0
The strong safety funnel ARNOLD V2.0 with lid and ball meets the highest requirements for safe disposal of hazardous liquid waste due to its combination of two independent safety mechanisms.
Brand: SCAT™ 317644
137.82 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
The safety funnel ARNOLD V2.0 with lid and ball valve is available for different container threads. In order to be able to produce in an environmentally friendly and economical way, we supply individual variants with a thread adapter, which has no influence on the functionality or safety of the funnel.
- Two safety features consist of a ball integrated in the funnel basin and a sealing ring incorporated in the hinged lid of the funnel, which prevent the escape of toxic or explosive gases
- Made of high quality HDPE material for maximum stability and durability
- Hinged lid with sealing ring for closure when the funnel is not in use
- Optimized sieve for catching larger particles and for depositing laboratory bottles
- Improved splash guard with integrated air channels to prevent bubble formation
- Ball valve with automatic opening mechanism during filling
- Individually shortenable safety lance for clean drainage of liquid waste
- All essential components are removable for easy cleaning
Specifications
200 mm (Funnel) | |
Not Autoclavable | |
Safe Disposal of Liquid and Volatile Wastes | |
Lance (220 mm), Lid, Ball Valve, Splash Guard, Removable Sieve | |
Safety Funnel |
HDPE | |
Blue, White | |
Disposal Containers | |
S51 |