missing translation for 'onlineSavingsMsg'
Learn More

SCAT™ Funnel ARNOLD, Ball-Valve

Product Code. 17172245 Shop All SCAT Europe Products
Click to view available options
Thread Size (Metric):
B53
B83
GL45
S50
S51
S55
S60/61
S65
S70/71
S90
S95
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17172245

Brand: SCAT™ 317647

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The strong safety funnel ARNOLD V2.0 with lid and ball meets the highest requirements for safe disposal of hazardous liquid waste due to its combination of two independent safety mechanisms.

The safety funnel ARNOLD V2.0 with lid and ball valve is available for different container threads. In order to be able to produce in an environmentally friendly and economical way, we supply individual variants with a thread adapter, which has no influence on the functionality or safety of the funnel.

  • Two safety features consist of a ball integrated in the funnel basin and a sealing ring incorporated in the hinged lid of the funnel, which prevent the escape of toxic or explosive gases
  • Made of high quality HDPE material for maximum stability and durability
  • Hinged lid with sealing ring for closure when the funnel is not in use
  • Optimized sieve for catching larger particles and for depositing laboratory bottles
  • Improved splash guard with integrated air channels to prevent bubble formation
  • Ball valve with automatic opening mechanism during filling
  • Individually shortenable safety lance for clean drainage of liquid waste
  • All essential components are removable for easy cleaning

Specifications

Diameter (Metric)Top 200 mm (Funnel)
Capacity (Metric) 1500 mL
Material HDPE
Height (Metric) 210 cm
Autoclavable Not Autoclavable
Color Blue, White
For Use With (Application) Safe Disposal of Liquid and Volatile Wastes
For Use With (Equipment) Disposal Containers
Includes Lance (220 mm), Lid, Ball Valve, Splash Guard, Removable Sieve
Thread Size (Metric) B83
Product Type Safety Funnel
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.