Learn More
SCAT™ Safety Funnel 'ARNOLD' with Ball and Lid, V2.0, HDPE-EC
The strong and electrically conductive safety funnel ARNOLD V2.0 with lid and ball meets the highest requirements for safe disposal of hazardous liquid waste due to its combination of two independent safety mechanisms.
Brand: SCAT™ 317629
Description
The safety funnel ARNOLD V2.0 with lid and ball valve is available for different container threads. In order to be able to produce in an environmentally friendly and economical way, we supply individual variants with a thread adapter, which has no influence on the functionality or safety of the funnel.
- Two safety features consist of a ball integrated in the funnel basin and a sealing ring incorporated in the hinged lid of the funnel, which prevent the escape of toxic or explosive gases
- Electrically conductive HDPE material with grounding connection to prevent flying sparks
- Hinged lid with sealing ring for closure when the funnel is not in use
- Optimized sieve for collecting larger particles and for depositing laboratory bottles
- Improved splash guard with integrated air channels to prevent bubble formation
- Ball valve with automatic opening mechanism during filling
- Individually shortenable safety lance for clean drainage of liquid waste
- All major components are removable for easy cleaning
Specifications
200 mm (Funnel) | |
Not Autoclavable | |
Safe Disposal of Liquid and Flammable Wastes | |
Lance (220 mm), Splash Guard, Removable Sieve, Grounding Cable, Lid, Ball Valve | |
7.9 in. (Funnel) |
HDPE-EC | |
Black | |
Disposal Containers | |
S50 | |
Safety Funnel |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.