missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fura-2, AM, cell permeant (1 mM Solution in Anhydrous DMSO)

Product Code. 10482492
Click to view available options
Quantity:
1 mL, 1 mL
Unit Size:
1mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10482492

Brand: Invitrogen™ F1225

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Fura-2, AM is a high affinity, intracellular calcium indicator that is ratiometric and UV light–excitable.

Fura-2, AM is a high affinity, intracellular calcium indicator that is ratiometric and UV light–excitable. This acetoxymethyl (AM) ester form is useful for noninvasive intracellular loading and is also available in 1 mg amounts (Cat. No. F-1201) and in special packagaing (F-1221). Cell-impermeant pentapotassium (F-1200) and pentasodium (F-6799) salt forms of the dye are available.

TRUSTED_SUSTAINABILITY

Specifications

Description Fura-2 AM, 1mL, Yellow, Liquid, 1001.86 Molecular Weight, C44H47N3O24 Molecular Formula, 189°C Boiling Point, 18.4°C Melting Point, 94°C Flash Point, Fluorescent Detection, –5 to –30°C Storage, Use with Fluorescence Microscope
Concentration Dimethylsulfoxide - 60 to 100%, 1 mM
Format Liquid
Content And Storage Store in freezer (-5°C to -30°C) and protect from light., -5°C to -30°C
For Use With (Application) For cell viability, proliferation and function, cellular imaging, pharma and biopharma, target and lead identification and validation, Cell Viability and Proliferation
Quantity 1 mL, 1 mL
Product Type Fura-2 AM, Fura-2
Detection Method Fluorescence, Fluorescent
For Use With (Equipment) Fluorescence Microscope, For fluorescence microscope
Dye Type Fluorescent Dye-Based
Color Yellow
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.