missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Fura-2, AM, cell permeant

Fura-2, AM, cell permeant

Brand:  Molecular Probes™ F1201

237.50 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.


 View more versions of this product

Product Code. 10308392

  • £257.00 / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Fura-2, AM is a high affinity, intracellular calcium indicator that is ratiometric and UV light—excitable. This acetoxymethyl (AM) ester form is useful for noninvasive intracellular loading and is also available in a DMSO solution. Cell-impermeant pentapotassium and pentasodium salt forms of the dye are available.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in freezer -5°C to -30°C and protect from light.
Cell Viability, Proliferation and Cellular Imaging
Calcium Indicator
1mg
Fluorescence
Fluorescence Microscope
Fluorescent Dye-Based
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.