Learn More
Molecular Probes™ Fura-2, Pentasodium Salt, cell impermeant
Fura-2, Pentasodium Salt, cell impermeant
Brand: Molecular Probes™ F6799
This item is not returnable.
View return policy
Description
Fura-2, pentasodium salt is an intracellular calcium indicator that is ratiometric and UV light—excitable. This water-soluble salt form is useful for intracellular loading by microinjection, infusion from patch pipette or uptake induced by our Influx pinocytic cell-loading reagent (I-14402).Specifications
Store in freezer -5°C to -30°C and protect from light. | |
Fluorescence Microscope, Microphotometer, Flow Cytometer | |
Fluorescent Dye-Based |
Fluorescence | |
Calcium Indicator | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.