missing translation for 'onlineSavingsMsg'
Learn More

Furan-3-boronic acid, 97%, Thermo Scientific Chemicals

Product Code. 10028811
Click to view available options
Quantity:
100 mg
500 mg
2.5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55552-70-0
Molecular Formula C4H5BO3
Molecular Weight (g/mol) 111.89
MDL Number MFCD01319007
InChI Key CYEFKCRAAGLNHW-UHFFFAOYSA-N
Synonym furan-3-boronic acid, 3-furanboronic acid, 3-furylboronic acid, furan-3-yl boronic acid, 3-furanylboronic acid, furan-3-ylboranediol, 3-furan boronic acid, furan-3-yl-3-boronic acid, furan-3-yl-boronic acid, boronic acid, 3-furanyl
PubChem CID 2734358
IUPAC Name furan-3-ylboronic acid
SMILES OB(O)C1=COC=C1

Specifications

Melting Point 126°C to 128°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Formula Weight 111.89
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Furan-3-boronic acid
compliance-icons
Product Identifier
  • Furan-3-boronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.