missing translation for 'onlineSavingsMsg'
Learn More
Furan, 99+%, stabilized, Thermo Scientific Chemicals Product Code.: 10676382

Furan, 99+%, stabilized, Thermo Scientific Chemicals

Product Code. 10676382
250 mL, Glass bottle
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10676382

Brand: Thermo Scientific Acros 181112500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-00-9, 128-37-0
Molecular Formula C4H4O
Molecular Weight (g/mol) 68.08
MDL Number MFCD00003222
InChI Key YLQBMQCUIZJEEH-UHFFFAOYSA-N
Synonym divinylene oxide, furfuran, tetrole, oxole, oxacyclopentadiene, furane, 1,4-epoxy-1,3-butadiene, axole, rcra waste number u124, furfuraneShow MoreShow Less
PubChem CID 8029
ChEBI CHEBI:35559
IUPAC Name furan
SMILES O1C=CC=C1

Specifications

Melting Point -85.6°C
Color Colorless to Yellow
Density 0.9360g/mL
Boiling Point 31.3°C
Flash Point −35°C
Infrared Spectrum Authentic
Assay Percent Range 0.05% max. (Furfural) (GC)
Packaging Glass bottle
Refractive Index 1.4197 to 1.4217
Quantity 250 mL
UN Number 2389
Beilstein 17, 27
Fieser 07,161; 09,227; 10,187; 13,173
Merck Index 15, 4325
Specific Gravity 0.936
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in alcohol,ether and aceton,hydrolysis in water
Stabilizer 0.025 to 0.04% BHT
Formula Weight 68.06
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Furan
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0453242 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number181110010,181112500,181110025
Lot NumberA0404504 Certificate TypeCertificate of Analysis Date26/07/2021 Catalog Number181110010,181112500,181110025
Lot NumberA0431259 Certificate TypeCertificate of Analysis Date16/06/2021 Catalog Number181110010,181112500,181110025
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Furan, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1B
  • Flammable liquid Category 1
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H350-May cause cancer.
  • H315-Causes skin irritation.
  • H341-Suspected of causing genetic defects.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H224-Extremely flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Furan, 99+%, stabilized, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.