missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Furegrelate Sodium Salt, ≥99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Förpackningsstorlek
10mg
50mg
5mg
Beskrivning
Inhibitor of thrombaxane synthetase
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifikationer
Specifikationer
| Quantity | 5 mg |
| Description | White to Off-white |
| Content And Storage | -30°C to -10°C |
| For Use With (Application) | Inhibitor of thrombaxane synthetase |
| Formula Weight | 275.20g/mol |
| Solubility | Soluble in water at approximately 15mg/ml |
| Product Type | Signal Transduction Reagent |
| Form | Crystalline solid |
| InChI Key | XBTIPIZROJAKOJ-UHFFFAOYSA-M |
| SMILES | [Na+].[O-]C(=O)C1=CC2=CC(CC3=CC=CN=C3)=CC=C2O1 |
| Visa mer |
RUO – Research Use Only
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering