missing translation for 'onlineSavingsMsg'
Learn More

Furfuryl Alcohol, 98%

Product Code. 10701155 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
250mL
This item is not returnable. View return policy

Product Code. 10701155

Brand: Thermo Scientific Acros 119790100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98-00-0 | C5H6O2 | 98.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-00-0
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.1
MDL Number MFCD00003252
InChI Key XPFVYQJUAUNWIW-UHFFFAOYSA-N
Synonym furfuryl alcohol, 2-furanmethanol, 2-furylmethanol, 2-furancarbinol, 2-furylcarbinol, furfuranol, 2-furanylmethanol, furfural alcohol, 2-furfuryl alcohol, 5-hydroxymethylfuran
PubChem CID 7361
ChEBI CHEBI:207496
IUPAC Name furan-2-ylmethanol
SMILES C1=COC(=C1)CO

Specifications

Melting Point -29°C
Color Yellow
Density 1.1300g/mL
Boiling Point 170°C
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic drum
Refractive Index 1.4850 to 1.488
Quantity 10 L
Beilstein 17, V,3, 338
Fieser 01,408
Merck Index 15, 4334
Specific Gravity 1.13
Solubility Information Solubility in water: miscible but unstable
Viscosity 5 mPa.s (25°C)
Formula Weight 98.1
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Furfuryl alcohol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.