missing translation for 'onlineSavingsMsg'
Learn More

Furfuryl alcohol, 98%

Product Code. 11484403
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 11484403

Brand: Thermo Scientific Alfa Aesar A10968.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98-00-0 | C5H6O2 | 98.101 g/mol

Furfuryl alcohol was used in the synthesis of continuous titanium carbide nanofibers/nanoribbon. Furfuryl alcohol is an efficient trapping agent for singlet oxygen determination in natural waters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Furfuryl alcohol was used in the synthesis of continuous titanium carbide nanofibers/nanoribbon. Furfuryl alcohol is an efficient trapping agent for singlet oxygen determination in natural waters.

Solubility
Soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep containers tightly closed in a cool, well-ventilated place. Keep in properly labeled containers. Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-00-0
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.101
MDL Number MFCD00003252
InChI Key XPFVYQJUAUNWIW-UHFFFAOYSA-N
Synonym furfuryl alcohol, 2-furanmethanol, 2-furylmethanol, 2-furancarbinol, 2-furylcarbinol, furfuranol, 2-furanylmethanol, furfural alcohol, 2-furfuryl alcohol, 5-hydroxymethylfuran
PubChem CID 7361
ChEBI CHEBI:207496
IUPAC Name furan-2-ylmethanol
SMILES C1=COC(=C1)CO

Specifications

Melting Point -31°C
Density 1.129
Boiling Point 170°C to 171°C
Flash Point 65°C (149°F)
Odor Mild
Refractive Index 1.487
Quantity 250 g
UN Number UN2874
Beilstein 106291
Merck Index 14,4305
Solubility Information Soluble in water.
Formula Weight 98.1
Percent Purity 98%
Chemical Name or Material Furfuryl alcohol
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.