missing translation for 'onlineSavingsMsg'
Learn More

Furfurylamine, 99%, Thermo Scientific Chemicals

Product Code. 11476856
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Furfurylamine is used as a water miscible solvent and as an intermediate in manufacturing pharmaceuticals like diuretics, antihypertensive, and antiseptic agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Furfurylamine is used as a water miscible solvent and as an intermediate in manufacturing pharmaceuticals like diuretics, antihypertensive, and antiseptic agents.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are strong oxidizing agents, air, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-89-0
Molecular Formula C5H7NO
Molecular Weight (g/mol) 97.117
MDL Number MFCD00003258
InChI Key DDRPCXLAQZKBJP-UHFFFAOYSA-N
Synonym furfurylamine, 2-furfurylamine, 2-furanmethylamine, 2-aminomethylfuran, 2-aminomethyl furan, 2-furanmethanamine, 2-furylmethylamine, 2-furylmethyl amine, usaf q-1, methylamine, 1-2-furyl
PubChem CID 3438
IUPAC Name furan-2-ylmethanamine
SMILES C1=COC(=C1)CN

Specifications

Melting Point -70°C
Density 1.05
pH 11.6
Boiling Point 145°C to 146°C
Flash Point 37°C (99°F)
Odor Amine-like
Refractive Index 1.49
Quantity 500 g
UN Number UN2526
Beilstein 1614
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 97.12
Percent Purity 99%
Chemical Name or Material Furfurylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Furfurylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Furfurylamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'