Learn More
Fursultiamine, MedChemExpress
MedChemExpress Fursultiamine is a vitamin B1 derivative, has anti-nociceptive and antineoplastic activity. Fursultiamine can be used for vitamin B1 deficiency, osteoarthritis (OA) and cancer research.
Brand: Medchem Express HY-B2082
This item is not returnable.
View return policy
Description
Fursultiamine is a vitamin B1 derivative, has anti-nociceptive and antineoplastic activity. Fursultiamine can be used for vitamin B1 deficiency, osteoarthritis (OA) and cancer research.Specifications
Fursultiamine | |
White | |
C17H26N4O3S2 | |
DMSO : 106.7 mg/mL (267.73 mM; Need ultrasonic and warming) | |
398.54 | |
99.86% | |
Solid |
804-30-8 | |
Research | |
5 mg | |
O=CN(CC1=CN=C(C)N=C1N)/C(C)=C(SSCC2OCCC2)\CCO | |
398.54 | |
Research | |
COVID-19-immunoregulation |