Learn More
Fursultiamine, MedChemExpress
MedChemExpress Fursultiamine is a vitamin B1 derivative, has anti-nociceptive and antineoplastic activity. Fursultiamine can be used for vitamin B1 deficiency, osteoarthritis (OA) and cancer research.
Brand:  Medchem Express HY-B2082
Encompass_Preferred
 View more versions of this product
Product Code. 16424647
  • £142.78 / 50mg
Explore more special offers
Description
Description
Fursultiamine is a vitamin B1 derivative, has anti-nociceptive and antineoplastic activity. Fursultiamine can be used for vitamin B1 deficiency, osteoarthritis (OA) and cancer research.
Specifications
Specifications
Fursultiamine
White
C17H26N4O3S2
DMSO : 106.7 mg/mL (267.73 mM; Need ultrasonic and warming)
398.54
99.86%
Solid
804-30-8
Research
50 mg
O=CN(CC1=CN=C(C)N=C1N)/C(C)=C(SSCC2OCCC2)\CCO
398.54
Research
COVID-19-immunoregulation