Learn More
VACUUBRAND™ Fuses Set For NT Diaphragm Pumps
The pigtail fuses are integrated into wires ((1), black and blue) inside the terminal box.To replace the fuses it is necessary to replace both wires completely(fixed with flat pin bushings(2))
Brand:  VACUUBRAND™ 20636542
Product Code. 11960142
  • £29.55 / Each
Estimated Shipment: 28-11-2024
to see stock.
Explore more special offers
Add to basket
Specifications
For NT Diaphragm Pumps
1 Set
Fuses Set