missing translation for 'onlineSavingsMsg'
Learn More

FUT8 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA565280

Product Code. 15988034

  • £428.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Immunogen sequence: MTDLYYLSQTD GAGDWREKEA KDLTELVQRR ITYLQNPKDC SKAKKLVCNI NKGCGYGCQL HHVVYCFMIA YGTQRTLILE SQNWRYATGG WETVFRP Highest antigen sequence identity to the following orthologs - mouse 99%, rat 99%.

Catalyzes the addition of fucose in alpha 1-6 linkage to the first GlcNAc residue, next to the peptide chains in N-glycans.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FUT8
Polyclonal
Unconjugated
FUT8
alpha (1,6) fucosyltransferase; alpha-(1,6)-fucosyltransferase; alpha-1,6-fucosyltransferase; alpha1-6FucT; EC 2.4.1.68; Fucosyltransferase 8; fucosyltransferase 8 (alpha (1,6) fucosyltransferase); FUT8; GDP-fucose--glycoprotein fucosyltransferase; GDP-L-Fuc:N-acetyl-beta-D-glucosaminide alpha1,6-fucosyltransferase; Glycoprotein 6-alpha-L-fucosyltransferase; MGC26465
Rabbit
Antigen affinity chromatography
RUO
2530
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.08 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9BYC5
FUT8
Recombinant Human FUT8
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only