Learn More
Molecular Probes™ FxCycle™ PI/RNase Staining Solution
FxCycle™ PI/RNase Staining Solution
Brand:  Molecular Probes™ F10797
119.70 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Product Code. 13433329
  • £130.00 / 100mL
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
FxCycle™ PI/RNase Staining Solution is used for flow cyotmetric analysis of DNA content in fixed cells. Propidium iodide (PI), a popular red-fluorescent stain, binds to DNA by intercalating between the bases with little or no sequence preference and with a stoichiometry of one dye per 4–5 base pairs of DNA. PI also binds to RNA, necessitating treatment with RNase to distinguish between RNA and DNA staining. This propidium iodide solution includes DNase-free RNase A and a permeablization reagent. It comes ready to use: simply add the staining solution to fixed cells, incubate, and acquire on a flow cytometer without washing.
View a selection guide for all products related to cell cycle analysis of fixed and live cells in flow cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 bottle FxCycle™ PI/RNase Staining Solution (100 mL). Store at room temperature, and protect from light.Store at room temperature
propidium iodide
Flow Cytometer
Fluorescence
FxCycle™
Staining Solution
535⁄ 617
Tube(s)
100 mL
Solution
Room Temperature
Nucleus