missing translation for 'onlineSavingsMsg'
Learn More

FYTTD1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen FYTTD1
Concentration 0.36 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16803045
View Documents
Proteintech
24560-1-AP-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16893035
View Documents
Proteintech
24560-1-AP-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FYTD1 required for mRNA export from the nucleus to the cytoplasm. This protein acts as an adapter that uses the BAT1/UAP56-NFX1 pathway to ensure efficient mRNA export and delivering to the nuclear pore. FYTD1 associates with spliced and unspliced mRNAs simultaneously with THOC4.
Specifications

Specifications

FYTTD1
Western Blot
Unconjugated
Rabbit
Human
Q96QD9
84248
FYTTD1 Fusion Protein Ag20051
Primary
-20°C
FYTTD1
0.36 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
FYTTD1, Protein 40 2 3, UAP56 interacting factor, UIF
FYTTD1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications